Regulatory Compliance
Refrigerator Capacity (Liters)
Refrigerator Capacity (cu ft)
Max Temperature - Refrigerator (° C)
Industry
North Sciences
North Sciences
North Sciences, an inTest company, designs and engineers dependable, low maintenance refrigerators and ultra-low temperature (ULT) freezers that meet a wide range of industry requirements. They are available with Traceable® data-loggers and the option to add TraceableLIVE® cloud monitoring service. North Sciences is one of a few companies in the world to deliver auto cascade refrigeration systems down to -100°C. Cole-Parmer is proud to add this reliable laboratory equipment to its product portfolio. These freezers are ideal for biomedical, pharmaceutical, life sciences and healthcare industries. North Sciences is an ISO 9001 registered company.