Product Type
Column Length (cm)
Column Length (mm)
Methodology
Industry
Chromaflex
Chromaflex
CHROMAFLEX columns are used in aqueous and organic chromatography. They contain PTFE-shielded o-rings which form a solvent-resistant, leak-free seal. We are a CHROMAFLEX authorized distributor.