Traceable’s research and development team embarked on a journey of customer focus groups and industry conversations to learn how clinics, labs, academic institutions, logistics companies, and other organizations use our temperature instruments, timers, and hygrometers. We used what we learned to update four of Traceable’s top-selling products to be even more user-friendly, economical, and higher performing than ever. All that and they’re still calibrated to NIST-traceable standards.
Traceable Refrigerator/Freezer Digital Thermometer with Probe
Be alerted to any issues affecting the contents of your fridge without even opening the door.
1Locate issues quickly and eliminate panic when the alarm Indicator bar lights up brightly to draw attention to an issue immediately. Once the trouble has been identified, the Buzzer Mute button allows you to focus on a resolution.
2Know about excursions BEFORE they happen with the pre-alert alarm light that glows orange when your fridge or freezer is within one degree of your alarm setting. This feature is a Traceable® Exclusive!
3Take readings quickly and with certainty. Triple display shows high, low, and current temperature simultaneously.
4 Pick the probe that fits your application from multiple probe options.
5Staff onboarding is a breeze with an enhanced, more intuitive user experience including large buttons, high-contrast text, and purposeful design.
6Maintain compliance. Includes NIST-traceable certificate from our A2LA-accredited (1750.01) calibration laboratory.
7 Reduce upkeep with boosted battery power and interior tech upgrades for higher performance.
Traceable Thermohygrometer with Clock
Monitor temperature, humidity, and time simultaneously, quickly. All three parameters are displayed on screen, all the time, so you can log conditions without putting down your clipboard.
1Features Min/Max memory, which is easily cleared at the beginning of each process.
2Staff onboarding is a breeze with an enhanced, more intuitive user experience including large buttons, high-contrast text, and purposeful design.
3Can be placed anywhere, whether magnetically mounted or set on the benchtop with built-in kick-stand.
4 Maintain compliance. Includes NIST-traceable certificate from our A2LA-accredited (1750.01) calibration laboratory.
5Reduce upkeep with boosted battery power and interior tech upgrades for higher performance.
Traceable Benchtop 3-Channel Timer with Alarm
Calibrated timer developed with busy clinics and clinical labs in mind. Always ready on your benchtop with three channels available for tracking simultaneous timing events.
1Antimicrobial coating is built in for a cleaner, more hygienic product.
2Maintain compliance. Includes NIST-traceable certificate from our A2LA-accredited (1750.01) calibration laboratory.
3With three channels to time multiple events, you can time various events at one time.
4 Large, tactile buttons that are easy to use when gloved.
5Features on-board memory function, time, date, and increase/decrease buttons for faster setup.
6Volume and buzzer timers are adjustable.
7Ergonomic design provides optimal viewing angle, easy use, and increased stability on bench.
Traceable Multitasking 4-Channel Timer with Clock
Calibrated timer and digital clock is always near-at-hand with its clip-on, magnet, benchtop configurations and adjustable viewing angle.
1Maintain compliance. Includes NIST-traceable certificate from our A2LA-accredited (1750.01) calibration laboratory.
2This timer steps in time with four channels to time multiple events at once.
3Never miss your cue with extra-loud alarms that sound for one full minute.
4Features on-board memory function, time, date, and increase/decrease buttons for faster setup.
5Antimicrobial coating is built in for a cleaner, more hygienic product.
6Large, tactile buttons that are easy to use when gloved.
7Ergonomic design provides optimal viewing angle, easy use, and increased stability on bench.