Reprinted with permission of THERMO
The purpose of this tutorial is to introduce basic concepts of centrifugation, including vocabulary, centrifuge and rotor types, separation techniques, and even gradient selection. For further details regarding centrifugation, please refer to the Sorvall® and Heraeus recommended reading the literature cited in the reference section.
Contents.
I. Introduction
II. Increasing the effect of gravity: the centrifuge
III. Types of centrifugal separation
IV. Rotor categories
V. Selection of centrifuge tubes
VI. Common centrifuge vocabulary and formulas
VII. Reference and suggested readings
Centrifugation is one of the most important and widely applied research techniques in biochemistry, cellular and molecular biology, and in medicine. Current research applications rely on isolation of cells, subcellular organelles, and macromolecules, often in high yields.
A centrifuge uses centrifugal force (g-force) to isolate suspended particles from their surrounding medium on either a batch or a continuous-flow basis. Applications for centrifugation are many and may include sedimentation of cells and viruses, separation of subcellular organelles, and isolation of macromolecules such as DNA, RNA, proteins, or lipids.
Many particles or cells in a liquid suspension, given time, will eventually settle at the bottom of a container due to gravity (1 x g). However, the length of time required for such separations is impractical. Other particles, extremely small in size, will not separate at all in solution, unless subjected to high centrifugal force. When a suspension is rotated at a certain speed or revolutions per minute (RPM), centrifugal force causes the particles to move radially away from the axis of rotation. The force on the particles (compared to gravity) is called Relative Centrifugal Force (RCF). For example, an RCF of 500 x g indicates that the centrifugal force applied is 500 times greater than Earth’s gravitational force. Table 1 illustrates common centrifuge classes and their applications.
Table 1. Classes of centrifuges and their applications
Table scrolls horizontally
| Centrifuge Classes | |||
| Low-speed | High-speed | Ultra/micro-ultra | |
| Maximum Speed (rpm x103) | 10 | 28 | 100/150 |
| Maximum RCF (x103) | 7 | 100 | 800/900 |
| Pelleting applications Bacteria | Yes | Yes | (Yes) |
| Animal and plant cells | Yes | Yes | (Yes) |
| Nuclei | Yes | Yes | (Yes) |
| Precipitates | Some | Most | (Yes) |
| Membrane fractions | Some | Some | Yes |
| Ribosomes/Polysomes | - | - | Yes |
| Macromolecules | - | - | Yes |
| Viruses | - | Most | Yes |
| Viruses | - | Most | Yes |
() = can be done but not usually used for this purpose.
Separation is achieved primarily based on the size of the particles in differential centrifugation. This type of separation is commonly used in simple pelleting and in obtaining partially-pure preparation of subcellular organelles and macromolecules. For the study of subcellular organelles, tissue or cells are first disrupted to release their internal contents. This crude disrupted cell mixture is referred to as a homogenate. During centrifugation of a cell homogenate, larger particles sediment faster than smaller ones and this provides the basis for obtaining crude organelle fractions by differential centrifugation. A cell homogenate can be centrifuged at a series of progressively higher g-forces and times to generate pellets of partially-purified organelles.
When a cell homogenate is centrifuged at 1000 x g for 10 minutes, unbroken cells and heavy nuclei pellet to the bottom of the tube. The supernatant can be further centrifuged at 10,000 x g for 20 minutes to pellet subcellular organelles of intermediate velocities such as mitochondria, lysosomes, and microbodies. Some of these sedimenting organelles can obtained in partial purity and are typically contaminated with other particles. Repeated washing of the pellets by resuspending in isotonic solvents and re-pelleting may result in removal of contaminants that are smaller in size (Figure 1). Obtaining partially-purified organelles by differential centrifugation serves as the preliminary step for further purification using other types of centrifugal separation (density gradient separation).
Density gradient centrifugation is the preferred method to purify subcellular organelles and macromolecules. Density gradients can be generated by placing layer after layer of gradient media (Table 2) such as sucrose in a tube with the heaviest layer at the bottom and the lightest at the top in either a discontinuous or continuous mode. The cell fraction to be separated is placed on top of the layer and centrifuged. Density gradient separation can be classified into two categories. 2a. Rate-zonal (size) separation. 2b. Isopycnic (density) separation.
Rate-zonal separation takes advantage of particle size and mass instead of particle density for sedimentation. Figure 2 illustrates a rate-zonal separation process and the criteria for successful rate-zonal separation. Examples of common applications include separation of cellular organelles such as endosomes or separation of proteins, such as antibodies. For instance, Antibody classes all have very similar densities, but different masses. Thus, separation based on mass will separate the different classes, whereas separation based on density will not be able to resolve these antibody classes.
Certain types of rotors are more applicable for this type of separation than others. Please See rotor categories (below) and Table 2.
In this type of separation, a particle of a particular density will sink during centrifugation until a position is reached where the density of the surrounding solution is exactly the same as the density of the particle. Once this quasi-equilibrium is reached, the length of centrifugation does not have any influence on the migration of the particle. A common example for this method is separation of nucleic acids in a CsCl gradient. Figure 3 illustrates the isopycnic separation and criteria for successful separation. A variety of gradient media can be used for isopycnic separations and their biological applications are listed in Table 2.
Table 2. Applications of density gradient media for isopycnic separations
Table scrolls horizontally
| Gradient media | Cells | Viruses | Organelles | Nucleoproteins | Macromolecules |
| Sugars (e.g. sucrose) | + | +++ | +++ | + | - |
| Polysaccharides (e.g. Percoll) | ++ | ++ | ++ | - | - |
| Colloidal silica (e.g. Percoll) | +++ | + | +++ | - | - |
| Iodinated media | ++++ | ++ | ++++ | +++ | + |
| Alkali metal salts (e.g. CsCI) | - | ++ | - | ++ | ++++ |
++++ excellent, +++ good, ++ good for some applications, + limited use, - unsatisfactory
Source: D. Rickwood, T.C. Ford, J. Steensgard (1994) Centrifugation essential data, John Wiley & Sons Ltd. U.K.
Rotors can be broadly classified into three common categories namely swinging-bucket rotors, fixed-angle rotors, and vertical rotors (Figure 4, Table 3). Note that each type of rotor has strengths and limitations depending on the type of separation.
Other rotors include continuous flow and elutriation rotors.
Table 3. Types of rotors and their applications
| Type of rotor | Pelleting | Rate-zonal Sedimentation | Isopycnic |
| Fixed-angle | Excellent | Limited | Variable* |
| Swinging-Bucket | Inefficient | Good | Good** |
| Vertical | NS | Good | Excellent |
| Zonal | NS | Excellent | Good |
NS = not suitable
*Good for macromolecules, poor for cells, and organelles
**Good for cells and organelles, caution needed if used with CsCI
In swinging bucket rotors, the sample tubes are loaded into individual buckets that hang vertically while the rotor is at rest. When the rotor begins to rotate the buckets swing out to a horizontal position (Figure 4). This rotor is particularly useful when samples are to be resolved in density gradients. The longer pathlength permits better separation of individual particle types from a mixture. However, this rotor is relatively inefficient for pelleting. Also, care must be taken to avoid “point loads” caused by spinning CsCl or other dense gradient materials that can precipitate.
In fixed-angle rotors, the sample tubes are held fixed at the angle of the rotor cavity. When the rotor begins to rotate, the solution in the tubes reorients (Figure 4). This rotor type is most commonly used for pelleting applications. Examples include pelleting bacteria, yeast, and other mammalian cells. It is also useful for isopycnic separations of macromolecules such as nucleic acids.
In vertical rotors, sample tubes are held in vertical position during rotation. This type of rotor is not suitable for pelleting applications but is most efficient for isopycnic (density) separations due to the short pathlength. Applications include plasmid DNA, RNA, and lipoprotein isolations.
Table 4 and Table 5 illustrate properties of centrifuge tubes and the proper rotors in which they should be used.
Table 4. Chemical Compatibility of Popular Tube Materials
| Tube Plastic Type | Clarity | Chemical Resistance* |
| Polypropylene (PP) | Opaque | Good |
| Polyallomer (PA) | Opaque | Good |
| Polycarbonate (PC) | Clear | Poor |
| Polyethylene terephtalate (PET) | Clear | Poor |
*For more information, please check our chemical resistance chart available on this site.
Selection of the appropriate centrifuge tube:
Major factor in selection of a tube (plastic) material:
To prolong tube life and avoid breakage or collapse:
Table 5. Tube Type and Rotor Compatibility
| Rotor Type | |||
| Tube Type | Fixed-angle | Swinging-bucket | Vertical |
| Thin wall open top | No | Yes | No |
| Thick wall open top | Yes | Yes | No |
| Thin wall sealed | No | Some tube types | Yes |
| Oak Ridge | Yes | No | No |
Old Rotor (Beckman® JA-10) - New Rotor (Sorvall® SLC-1500)
T2 = 20 min; K2 = 3610; T1 = ( ? ) min; K1 = 1676
Old Pelleting Time = 20 min - New Pelleting Time = 9.2 min
Disclaimer: Cole-Parmer products are not approved or intended for, and should not be used for medical, clinical, surgical or other patient-oriented applications.