Environmental Express® StableWeigh TDS Weighing Vessel Start-Up Kit
Pre-treated, pre-weighed, and disposable for your convenience
  • This product complies with the following methods: SM 2540, ASTM D5907, BS EN 15216:2007
  • Vessels and filters are washed, dried, weighed, and individually marked
  • Save time — cools faster, requires fewer drying and weighing cycles
  • Disposable to eliminate cross-contamination
These weighing vessels reduce preparation time, perfect for busy labs analyzing a lot of samples. Each vessel complies with the following methods: SM 2540, ASTM D5907, BS EN 15216:2007. Each vessel is treated to conform with ASTM D5907 Standard Test Methods for Filterable Matter (Total Dissolved Solids) and Nonfilterable Matter (Total Suspended Solids) in Water, then individually printed with the final weight. The material is lightweight, inert to water or wastewater components, and heat resistant. Vessels cool quickly decreasing wait times between heating and weighing.
Choose the startup kit to begin processing samples immediately. The kit contains enough vessels and filters for 100 samples, a weighing stand to keep your vessel stable on the balance pan, a 5-place carrier/oven rack, a 6-place filling station to make transferring sample to vessel quicker and easier, and a static control system.
£4,490.70GBP / Each
£5,388.84GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
StableWeigh Start-Up Kit w/ 6-place Filling Station, 100/pk
£4,490.70GBP / Each
£5,388.84GBP (incl VAT)