Dynalon Disposable Polystyrene Cuvette for Fluorometry
Precision – with convenience!
  • Constructed of durable polystyrene
Use these economical, single-use cuvettes to obtain consistent spectrophotometric data. Available in standard, semi-micro, and fluorometry configurations. Use polystyrene cuvettes for visible spectral analysis.
£174.85GBP / Pkg of 500
£209.82GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
Dynalon 202295 Disposable Visible Cuvettes, polystyrene; 4.5 mL capacity, 500/pack 0634360 Mfr # 202295
COMPARE
Polystyrene 4.5 10
£174.85GBP / Pkg of 500
£209.82GBP (incl VAT)