The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.
Supco SL300TH Mini Temperature and Humidity Data Logger
Representative image only.
Supco SL300TH Mini Temperature and Humidity Data Logger
Supco | Mfr: SL300TH| Item: 2303972
Provide consice measurements for critical environmental conditions
£170.29GBP / Each
£204.35GBP (incl VAT)
for an additional
£252.21GBP / Each
£302.65GBP (incl VAT)
for an additional
£334.50GBP / Each
£401.40GBP (incl VAT)
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
£170.29GBP / Each
£204.35GBP (incl VAT)
Buy It Together
Current Item
Supco SL300TH Mini Temperature and Humidity Data Logger
Supco SL300TH Mini Temperature and Humidity Data Logger
2303972
£170.29GBP / Each
£204.35GBP (incl VAT)
+
InnoCal NIST-Traceable Calibration; Humidity and Temperature, High-Accuracy
1703020
£252.21GBP / Each
£302.65GBP (incl VAT)
=
Total Price 422.50 ADD 2 ITEMS TO CART
Product Description
Includes
software, 6 ft USB cable, and one 3 V lithium battery.
Key Features
  • Includes easy-to-use user configurable software
  • Units accurate to ±1°F, (±0.5°C)
  • 1 year warranty
More About this Item
These mini dataloggers are ideal for recording temperature and humidity conditions in labs, food storage facilities, warehouses, refrigerators, freezers, museums, greenhouses and more.
Features include high/low alarms, PC, delayed start or pushbutton control, user selectable stop on memory full, or memory rollover for continuous recording. Included software allows you to view graph and text simultaneously and provides the ability to calibrate dataloggers.
Units provide feature a standard USB interface, and includes software that is Windows® 98SE, ME, 2000, XP and above compatible.
1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature and humidity
  • Max Temperature (° F)160
  • Min Temperature (° F)-40
  • Max Temperature (° C)70
  • Min Temperature (° C)-40
  • Temperature Accuracy (° F)±1
  • Temperature Accuracy (° C)±0.5
  • Max Humidity RH (RH)99
  • Min Humidity RH (RH)0
  • Humidity Resolution (%)0.01
  • Humidity AccuracyRH Accuracy: ±2 from 10 to 90
  • Memory Size43,344 data points (temperature only); 21,672 data points (temperature, humidity, and dew point)
  • Battery Type1 3.6V Lithium (included)
  • Length (in)1/2
  • Height (in)3
  • Width (in)1/2
  • DescriptionMini Temperature and Hunidity Data Logger