Scienceware F37805-0004 4 liter Churn Sample Splitter
Representative image only.
Scienceware F37805-0004 4 liter Churn Sample Splitter
Scienceware | Mfr: F37805-0004| Item: UY-99160-00
Mix and divide a water sample into subsamples for testing
£743.60GBP / Each
£892.32GBP (incl VAT)
£743.60GBP / Each
£892.32GBP (incl VAT)
Product Description
Key Features
  • Polyethylene vessel with polypropylene spigot
  • Swing handle included for ease of transportation
More About this Item
Use the sample splitter to efficiently composite representative cross-section samples of water-sediment mixtures from rivers and streams. Composite samples are homogenized and then split into representative subsamples using the churn splitter. The spring-loaded spigot drains the subsamples into containers for water quality analysis.
Specifications
  • MaterialPolyethylene, polypropylene
  • Sample Volume4
  • Height (in)12
  • Diameter (in)6.5
  • Description4 liter Churn Sample Splitter