NSI Lab Solutions QCI-171 Multi-Analyte Solids Certified Reference Material, 1 L; 4/PK

Representative image only.

NSI Lab Solutions QCI-171 Multi-Analyte Solids Certified Reference Material, 1 L; 4/PK

NSI Lab Solutions | Item: 0500510

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Supplied ready-to-use—just shake and pour!
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • Multi-analyte QC standard to QA 6 solids analyses including fixed, volatile, and suspended solids
  • Actual concentrations may vary slightly lot to lot
  • Certified stable for up to 3 years from manufacture
  • A NIST Traceable Certified Reference Material

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

A whole volume multi-analyte standard for quality control analysis for 6 solids parameters. Each 100 mL aliquot contains:
Total Solids: 350 mg
Total Dissolved Solids: 250 mg
Total Suspended Solids: 100 mg
Volatile Suspended Solids: 31 mg
Fixed Suspended Solids: 70 mg
Fixed Dissolved Solids: 215 mg

Specifications
  • Element/IonTotal Suspended Solids, Total Dissolved Solids, Total Solids
  • FormLiquid
  • Volume (mL)1000
  • Packaging4/PK
  • Storage SymbolStore between 15 and 30°C
  • Shelf Life (months)36
  • DescriptionMulti-Analyte Solids Certified Reference Material, 1 L; 4/PK
Documents & Certificates
Reviews
Q&A