NSI Lab Solutions QCI-123 Total Chlorine Accuracy Check Snip™ CRM, 1.00 mg/L, 1.5 mL; 25/PK

Representative image only.

NSI Lab Solutions QCI-123 Total Chlorine Accuracy Check Snip™ CRM, 1.00 mg/L, 1.5 mL; 25/PK

NSI Lab Solutions | Item: 0500517

Validate analytical methods and evaluate the quality of your results with premeasured certified reference materials

Call For Price

Call For Price

Buy It Together

Current Item

NSI Lab Solutions QCI-123 Total Chlorine Accuracy Check Snip™ CRM, 1.00 mg/L, 1.5 mL; 25/PK
NSI Lab Solutions QCI-123 Total Chlorine Accuracy Check Snip™ CRM, 1.00 mg/L, 1.5 mL; 25/PK
0500517

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • For colorimetric total and free chlorine analysis
  • Certified stable for up to 2 years from manufacture
  • Each tube yields 25 mL of 1.00 ppm (mg/L) standard
  • 25 x 1.5 mL Snips per pack

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Using a Snip is a Snap! To use, snip open the PTFE tube and pour contents directly into a 25 mL cuvette then bring to volume with reagent. Each Snip yields 1.00 ppm chlorine working standard.

Specifications
  • Concentration1.00 mg/L
  • Element/IonTotal and Free Chlorine
  • FormLiquid
  • Volume (mL)1.5
  • Packaging25/PK
  • Storage SymbolRefrigerate between 2 and 8 °C
  • CAS Number7681-52-9
  • Shelf Life (months)24
  • DescriptionTotal Chlorine Accuracy Check Snip™ CRM, 1.00 mg/L, 1.5 mL; 25/PK
Documents & Certificates
Reviews
Q&A