NSI Lab Solutions QCI-118 Ultra-Low Chlorine Accuracy Check Snip™ CRM, 0.100 mg/L, 1.5 mL; 25/PK

Representative image only.

NSI Lab Solutions QCI-118 Ultra-Low Chlorine Accuracy Check Snip™ CRM, 0.100 mg/L, 1.5 mL; 25/PK

NSI Lab Solutions | Item: 0500516

Validate analytical methods and evaluate the quality of your results with premeasured certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • For colorimetric total and free chlorine analysis
  • Certified stable for up to 2 years from manufacture
  • Each tube yields 1000 mL of 0.100 ppm (mg/L) standard
  • 25 x 1.5 mL Snips per pack

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Using a Snip is a Snap! To use, snip open the PTFE tube and dilute contents to 1000 mL in reagent water. Each Snip yields 0.100 ppm chlorine working standard.

Specifications
  • Concentration0.100 mg/L
  • Element/IonTotal and Free Chlorine
  • FormLiquid
  • Volume (mL)1.5
  • Packaging25/PK
  • Storage SymbolRefrigerate between 2 and 8 °C
  • CAS Number7681-52-9
  • Shelf Life (months)24
  • DescriptionUltra-Low Chlorine Accuracy Check Snip™ CRM, 0.100 mg/L, 1.5 mL; 25/PK
Documents & Certificates
Reviews
Q&A