NSI Lab Solutions QCI-081SNIP GGA Concentrate Snip™ Certified Reference Material, 300 mg/L, 6 mL; 25/PK

Representative image only.

NSI Lab Solutions QCI-081SNIP GGA Concentrate Snip™ Certified Reference Material, 300 mg/L, 6 mL; 25/PK

NSI Lab Solutions | Item: 0500498

Validate analytical methods and evaluate the quality of your results with premeasured certified reference materials

Call For Price

Call For Price

Buy It Together

Current Item

NSI Lab Solutions QCI-081SNIP GGA Concentrate Snip™ Certified Reference Material, 300 mg/L, 6 mL; 25/PK
NSI Lab Solutions QCI-081SNIP GGA Concentrate Snip™ Certified Reference Material, 300 mg/L, 6 mL; 25/PK
0500498

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • Convenient, easy to use and accurate single-use tube
  • No measurement required—pour directly into a 300 mL BOD Bottle
  • Certified stable for up to 3 years from manufacture
  • 25 x 6 mL Snips per pack

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Using a Snip is a Snap! Each tube delivers exactly 300 ppm (mg/L) of NIST traceable GGA. To use, snip open the PTFE tube and pour contents directly into a 300-mL BOD bottle.

Specifications
  • Concentration300 mg/L
  • Element/IonGlucose-Glutamic Acid (GGA)
  • FormLiquid
  • Volume (mL)6
  • Packaging25/PK
  • Storage SymbolStore between 15 and 30 °C
  • CAS Number56-86-0, 50-99-7
  • Shelf Life (months)36
  • DescriptionGGA Concentrate Snip™ Certified Reference Material, 300 mg/L, 6 mL; 25/PK
Documents & Certificates
Reviews
Q&A