Fytertech Spilfyter Wastewater Spill Classifier Kit; 6 Tests/Kit
Representative image only.
Fytertech Spilfyter Wastewater Spill Classifier Kit; 6 Tests/Kit
Fytertech | Mfr: 580001| Item: 8642302
Classify unknown spills in 30 seconds
£112.83GBP / Each
£135.40GBP (incl VAT)
£112.83GBP / Each
£135.40GBP (incl VAT)
Buy It Together
Current Item
Fytertech Spilfyter Wastewater Spill Classifier Kit; 6 Tests/Kit
Fytertech Spilfyter Wastewater Spill Classifier Kit; 6 Tests/Kit
8642302
£112.83GBP / Each
£135.40GBP (incl VAT)
+
Fytertech Spilfyter Chemical test strips, case of 50
8642350
£785.66GBP / Case of 50
£942.79GBP (incl VAT)
+
Fytertech Spilfyter Wastewater Reference Chart
8642356
£27.13GBP / Each
£32.56GBP (incl VAT)
=
Total Price 925.62 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Kit includes six test strips, one bobbin of thread, one color reference chart, and one plastic tube with cap. Additional test strips and charts may also be ordered separately.
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid.
Wastewater classifier kit tests for acid or base, lead, fluoride, nitrate, nitrite, and petroleum/organic products.
Specifications
  • DescriptionWastewater Spill Classifier Kit; 6 Tests/Kit