NPS Spilfyter Hazmat Yellow Drum Top Spill Pad, Heavy-Weight, 22" diameter, 25/Case

Representative image only.
NPS Mfr # S2-76 – Item # WZ-86423-72
Coverstock uses spunbond material for excellent durability and virtually no linting

PRODUCT OPTIONS

£164.51GBP / Case of 25
£197.41GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Absorption14 gal. (53 L)
  • OD (in)22
  • Width (in)22
  • Width (cm)56
  • ColorYellow
  • DescriptionHazmat Yellow Drum Top Spill Pad, Heavy-Weight, 22\" diameter, 25/Case

More About this Item

Hazmat sorbents are specifically designed to absorb aggressive fluids such as acids and bases. Ideal for laboratory use.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat