Fytertech Spilfyter Grab & Go Universal Bucket Spill Kit, Absorbs 4 gallons
Representative image only.
Fytertech Spilfyter Grab & Go Universal Bucket Spill Kit, Absorbs 4 gallons
Fytertech | Mfr: 455304| Item: 8642412
Designed for portability and quick response to small and medium spills
£97.90GBP / Each
£117.48GBP (incl VAT)
£97.90GBP / Each
£117.48GBP (incl VAT)
Product Description
Includes
Kit includes four 3" x 4' socks, three 16" x 18" pads, one pair of nitrile gloves, one pair of goggles, and one 5-gallon plastic bucket with snap-on lid.
Key Features
  • Snap-on lid protects contents
More About this Item
Universal sorbents are ideal for environments where there are many different liquids including cutting oils, lubricants, and coolants.
Specifications
  • Absorption4 gal. (16 L)
  • DescriptionUniversal Bucket Spill Kit, Absorbs 4 gallons