Fytertech Spilfyter Grab & Go Acid Bucket Spill Kit, Liquid Formula
Representative image only.
Fytertech Spilfyter Grab & Go Acid Bucket Spill Kit, Liquid Formula
Fytertech | Mfr: 405104| Item: 8642452
Kit contents are packed in a 5-gallon bucket for easy portability and disposal of materials
£213.43GBP / Each
£256.12GBP (incl VAT)
£213.43GBP / Each
£256.12GBP (incl VAT)
Product Description
Includes
Three 3" x 4' hazmat socks, ten 12" x 12" hazmat pads, one gallon liquid acid neutralizer, two chemical classifier strips, one chemical classifier chart, pair of nitrile gloves, pair of goggles, disposal bag with twist tie, 5-gallon plastic bucket.
Key Features
  • Use chemical classifier to detect the presence of other chemicals
More About this Item
Grab & Go buckets contain everything you need to quickly clean-up and dispose of a spill. Kit includes materials to neutralize acids.
Specifications
  • Absorption3 gal. (12 L)
  • DescriptionAcid Bucket Spill Kit, Liquid Formula