Fytertech Spilfyter Chemical Reference Chart
Representative image only.
Fytertech Spilfyter Chemical Reference Chart
Fytertech | Mfr: 577777| Item: 8642354
Classify unknown spills in 30 seconds
£9.79GBP / Each
£11.75GBP (incl VAT)
£9.79GBP / Each
£11.75GBP (incl VAT)
Buy It Together
Current Item
Fytertech Spilfyter Chemical Reference Chart
Fytertech Spilfyter Chemical Reference Chart
8642354
£9.79GBP / Each
£11.75GBP (incl VAT)
+ =
Total Price 347.04 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Convenient strips change color to identify risk type and classify spill for proper treatment. Compare color indicator to laminated, reusable reference chart to classify unknown liquid. Chemical classifier kit tests for acid or base, iodine, oxidizers, chlorine, fluoride, bromine and petroleum/-organic products. Wastewater classifier kit tests for acid/base, petroleum/organic solvent, hydrogen sulfide, nitrite, nitrate, and fluoride risks.
Replacement reference chart for Chemical Classifier Kit.
Specifications
  • DescriptionChemical Reference Chart