The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.

Environmental Express PC 2000 Benchtop Meter Kit

Representative image only.

Environmental Express PC 2000 Benchtop Meter Kit

Environmental Express | Item: 3541965

Developed using extensive feedback from laboratory staff, our new Environmental Express 2000 series meters are assembled in various configurations for measuring pH/mV, EC, Ion, DO, BOD parameters

£1,596.21GBP / Each
£1,915.45GBP (incl VAT)

£1,596.21GBP / Each
£1,915.45GBP (incl VAT)

Buy It Together

Current Item

Environmental Express PC 2000 Benchtop Meter Kit
Environmental Express PC 2000 Benchtop Meter Kit
3541965

£1,596.21GBP / Each
£1,915.45GBP (incl VAT)
+ +
Environmental Express 2000 All-in-One pH Electrode
3541952

£159.29GBP / Each
£191.15GBP (incl VAT)
+
Environmental Express pH 2000 Benchtop Meter Kit
3541954

£958.23GBP / Each
£1,149.88GBP (incl VAT)
+
Environmental Express Oakton Ion Selective Electrode (Electrode type: Sodium)
3581242

£219.48GBP / Each
£263.38GBP (incl VAT)
=
Total Price 3917.10 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Electrode holder, universal power adapter with six plugs, 35419-52 All-In-One pH electrode 2000, 35419-47 1500 / 2000 conductivity probe, 3 pH buffer solutions (4.01, 7.00, 10.01), and 4 conductivity standards (84 µS, 1413 µS, 12.88 mS, 111.8 mS).

Key Features

  • Various configurations available to meet the demands of measuring pH/mV, EC, Ion, PC, PD, DO, BOD parameters
  • Meter Kit formats include probes, 35419-54, -60, -65, -74, -78
  • Automatic record retention and accessibility for continuous compliance with regulatory requirements
  • 0.01, 0.001 resolution for routine or research applications where acute accuracy is critical
  • Large displays with a small footprint, ideal for labs with limited bench space
  • Automatic functions like buffer recognition and auto calibration for ease of use at any skill level

More About this Item

Environmental Express provides a diverse range of water quality analyzers and electrodes that are designed specifically for laboratory applications with compliance in mind. Our multi-parameter benchtops include functionality to meet the highly specific and/or broad ranging needs of wastewater, drinking water, food and beverage, chemical and pharmaceutical testing labs.

These benchtop meters offer a high-capacity internal memory that can capture and store 2000 data points with the time and date. They have a time-based auto data log with GLP compliant real time clocks. This data can be exported into CSV, Excel, or PDF formats via USB cable and use of complimentary software. A password access protection function secures the meter setup mode from unauthorized access. An adjustable shutoff time up to 30 minutes turns the meter off when idle.

Specifications
  • Meter TypepH/ORP/Ion/Conductivity/Resistivity/TDS/Salinity/Temp (°C/°F)
  • Display type5” Custom LCD with backlight and 320 segments
  • Connections2 x BNC, 2 x phono (ATC), DC sockets
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.1 / 0.01 / 0.001 pH
  • pH Accuracy± 0.003 pH
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 mV
  • mV Accuracy± 0.2 mV
  • Min ISE0.000 µg/L
  • Max ISE9999 g/L
  • ISE Resolution4 Significant digits
  • ISE Accuracy± 0.3% full scale or ± 0.2 mV, whichever is higher
  • Min Conductivity0.000 µS/cm (k = 0.1)
  • Max Conductivity2 S/cm (k=10)
  • Conductivity Resolution0.05% full scale
  • Conductivity Accuracy± 0.6% full scale; ± 1.5% full scale > 18.0 mS/cm
  • Min TDS0.01 mg/L (ppm)
  • Max TDS100 g/L (ppt)
  • TDS Resolution0.01, 0.1, 1 mg/L - g/L (ppm - ppt)
  • TDS Accuracy± 0.1% full scale
  • Min Salinity0.00 ppt / 0.00%
  • Max Salinity100.0 ppt / 10.00%
  • Salinity Resolution0.1 ppt / 0.01%
  • Salinity Accuracy± 0.2% full scale
  • Min Resistivity0.000 Ω•cm
  • Max Resistivity20.0 MΩ•cm
  • Resistivity Resolution0.5% full scale
  • Resistivity Accuracy± 0.6% full scale; ± 1.5% full scale > 1.80 MΩ•cm
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1 °C / °F
  • Temperature Accuracy± 0.5 °C / ± 0.9 °F
  • OutputPhono jack (USB/RS-232C)
  • Data Logging (points)2000
  • Power (VAC)100 to 240
  • Power (Hz)50-60 Hz
  • Length (cm)15.5
  • Width (cm)15
  • Height (cm)6.7
  • Width (in)5 29/32
  • Length (in)6 7/64
  • Height (in)2 41/64
  • DescriptionPC 2000 Benchtop Meter Kit
  • Warranty3 years
Reviews
Q&A