This item is now discontinued. Please browse similar items
Extech LCR205 SMD Componenet Fixture for LCR Meter
Representative image only.
Extech Mfr # LCR205 – Item # WZ-20043-02
Optimize performance of your Extech meters
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSMD Componenet Fixture for LCR Meter
  • Warranty6 Months
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Models 20043-01, -02, -03, 23402-20, and -05 are all compatible with the Extech Passive Component LCR meter 20043-00.
Models 20045-59, 64, and -65 are compatible with the Extech MS6000 Series Two-Channel Digital Storage Oscilloscopes 20045-56 and -57.
Meters sold separately.