Thermocouple Meters
The product searched is discontinued, please browse below for replacements.
Thermocouple Meters
Thermocouple meters come with different pairing of materials allowing for a very wide range of applications. The different compositions are given in thermocouple types (letter names) which are standardized across the industry. The most common types (calibrations) are J, K, T, and E. Although the thermocouple calibration dictates the temperature range, the maximum range is also limited by the diameter of the thermocouple wire. Use the following criteria to choose the best thermocouple instrument type for your applications: temperature range (probe calibration type), chemical resistance of the thermocouple, shealth material (such as stainless steel), abrasion and vibration resistance, and probe length and diameter.
APPLY FILTERS
filter byclear all
27 RESULTS
SORT BY
Showing 1 - 24 of 27
2>