Understanding |
|
Conductivity and resistivity are both measures of the ability of a fluid to conduct electrical current. Conductivity is simply the reciprocal of resistivity: conductivity = 1/resistivity (See Table 1). In practice, conductivity units are typically used when referring to water ranging from drinking water to sea water, while resistivity units are reserved for ultra pure water such as deionized or reverse-osmosis water.
Table 1
| Conductivity | Resistivity |
| 0.01 µS | 100 MΩ |
| 0.055 µS | 18.0 MΩ |
| 0.1 µS | 10 MΩ |
| 1 µS | 1 MΩ |
| 10 µS | 0.1 MΩ |
| 100 µS | 0.01 MΩ |
| 1 mS | 1 kΩ |
The unit of conductivity is the Siemen (S). A millisiemen (mS) = 1/1,000 S, a microsiemen (µS) = 1/1,000,000 S. Conductivity is also referred to as electrical conductivity (EC) or specific conductance.
Resistivity units are expressed in Ohms (Ω). A kilo ohm (kΩ) = 1,000 Ω, a megaohm (MΩ) = 1,000,000 Ω.
Since conductivity and resistivity relate to an area between which current is measured it is common to see the units expressed per volume such as MΩ-cm or µS-cm or umhos-cm. Few realize that "mho" is ohm spelled backwards to indicate the reverse relationship to Ohm's law! A "mho" is equivalent to a Siemen and are used interchangeably.
Conductivity is greatly influenced by temperature. Most fluids increase in conductivity as temperature increases. Most ionic solutions will increase about 2% for each 1°C increase. Unfortunately, this temperature coefficient (TC) is not linear. In the case of high resistance water it can be closer to 5% or so per °C.
Many instruments adjust the conductivity value based on a TC and display a value that is said to be corrected or normalized to 25°C. The meter will automatically make corrections to the reading and display a value as if the sample was 25°C, no matter what the actual temperature is. Some instruments use a fixed TC of 2.0% per °C. Let us consider a meter that uses 2.0% TC to measure a 1413 µS standard at 25°C (77°F). If the standard is warmed to 30°C (86°F), the meter applies a correction of 5 degrees x 0.02% x 1413 µS = 141.3. Without correction (0.0% TC) the actual value of a 1413 µS standard of KCl at 30°C (86°F) is 1548 µS. As the meter corrects for temperature, it displays a value of 1548 minµS 141.3 = 1407 µS. When the sample cools to 25°C, it will again read 1413 µS as no correction is applied. Although conductivity cell response is immediate, temperature corrected values will fluctuate as the temperature measurement stabilizes.
| More Details or Order Online: Oakton® CON 6+ Handheld Conductivity Meters Oakton®Waterproof CON 610 Conductivity/TDS/Salinity/Resistivity Meters |
TC labels on conductivity calibration standards often provide a temperature table listing conductivity values at different temperatures. Conductivity meters with a fixed TC should be calibrated to the conductivity value at the meters normalization temperature, usually 25°C. Calibration to values other than the normalization temperature would only be appropriate if the meter did not utilize temperature correction, or if the TC was adjusted to 0.0%. As a general rule its always best to calibrate and measure as close to 25°C as possible when a TC is applied. Recording the temperature during calibration and measurement is good practice.
Another feature of advanced meters is a selectable normalization temperature. This allows temperature compensated readings to be adjusted either to 25°C (77°F) or another value, usually 20°C (68°F). The advantage here is that 20°C (68°F) is often closer to the actual sample temperatures than 25°C (77°F). When using a normalization temperature other than 25°C it is important to calibrate to the appropriate value of the conductivity standard at the specified normalization temperature. For example, a 1413 µS standard at 25°C should be calibrated to its value at 20°C which is 1278 µS.
While emphasis is given to the conductivity accuracy, it is important not to neglect the temperature accuracy. Although temperature is directly related to conductivity measurement it is often overlooked. Meter temperature accuracy should be verified and calibrated if necessary prior to conductivity calibration.
Its best to think of cell constant as the "cell efficiency". It's a factor that the meter uses to make the standard value agree with the measured value. Conductivity cells are offered with nominal values such as k=1.0. In reality, the effective cell constant may deviate several percent of the nominal value and is determined only after calibration. A nominal 1.0 cell may have a 0.97 constant after calibration and correction by the meter which had to make a 3% correction to the calibration value. Each time the meter and cell are recalibrated, this cell efficiency may change. A cell can change slowly over time from physical changes taking place such as chemical oxidization, scratches, coating, bending, stripping, etc.
It is important to choose a cell constant based on the anticipated measuring range (see table below). If a cell constant other than 1.0 is required, a meter with a selectable cell constant will be necessary.
| Cell Constant | Optimal Range |
| 0.01 µS | <1 µS |
| 0.1 µS | 0.5 to 200 µS |
| 1.0 µS | 10 to 2,000 µS |
| 10 µS | 1 to 200 mS |
It is also necessary to choose between a 2-cell or a 4-cell units. Skipping much of theory, 4-cell units are more expensive but considered better as they resist polarization effects and fouling, however for clean water applications, this advantage may be small.
High resistivity calibration standards are not commercially feasible. Precision resistors can be used in place of the conductivity/resistivity probe to verify response and accuracy meter, but this practice will not account for differences in efficiency of the sensing cells or bands in individual probes.
Conductivity standards in the 10 to 100 µS range are available commercially. Prolonged exposure to air as well as contamination from glassware and the cell itself can significantly increase the value of your standard. Individual single-use pouches are often preferred over bottled standards, as they do not have to be poured into a sample container and are not subject to contamination through reuse. It is vital to maintain a clean cell at all times and use good laboratory techniques for accurate and reproducible results.
Meters that use total dissolved (ionic) solids (TDS) measure conductivity and multiply the reading by a fixed or adjustable "TDS factor" to determine TDS. TDS values are usually expressed as parts per million (ppm) or ppt (parts per thousand). There are many limitations when using TDS. First, the TDS factor used is salt specific so if there are multiple or unknown salts in solution, its nearly impossible to determine the correct factor to use. Second, since ionic concentrations are not linear, the TDS factor changes with concentration. TDS values are generally not considered with low conductivity values.