Spex CLMS-SET Claritas PPT® Grade ICP-MS 7 Multi-Element Solution Standards Set with Mercury

Representative image only.

Spex CLMS-SET Claritas PPT® Grade ICP-MS 7 Multi-Element Solution Standards Set with Mercury

Spex | Item: 0456888

Multi-element standards for ICP and ICP-MS instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Calibrate ICP-MS instrumentation prior to analysis
  • Designed to contain virtually every element in the mass spectrum for concentration verification checks
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance

More About this Item

Spex CertiPrep® Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability. Choose from the ICP-MS set of 7 multi-element solutions with or without mercury.

Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-element solution set contains 7 solutions.


  • Solution 1 is a 125 mL bottle of DI water blank. This is available individually as part number 04567-57.
  • Solution 2 is a 125 mL bottle of Hydrochloric Acid blank. This is available individually as part number 04567-53.
  • Solution 3 is a 125 mL bottle of Nitric Acid blank. This is available individually as part number 04567-55.
  • Solution 4 is a 125 mL bottle of Multi-Element Solution 1 which contains 10 µg/mL (10 ppm) of Cerium, Dysprosium, Erbium, Europium, Gadolinium, Holmium, Lanthanum, Lutetium, Neodymium, Praseodymium, Samarium, Scandium, Terbium, Thorium, Thulium, Ytterbium, and Yttrium. This is available individually as part number 04568-80.
  • Solution 5 is a 125 mL bottle of Multi-Element Solution 2 which contains 10 µg/mL (10 ppm) of Aluminum, Arsenic, Barium, Beryllium, Bismuth, Cadmium, Calcium, Cesium, Chromium, Cobalt, Copper, Gallium, Indium, Iron, Lead, Lithium, Magnesium, Manganese, Mercury, Nickel, Potassium, Rubidium, Selenium, Silver, Sodium, Strontium, Thallium, Uranium, Vanadium, and Zinc. This is available individually as part number 04568-81.
  • Solution 6 is a 125 mL bottle of Multi-Element Solution 3 which contains 10 µg/mL (10 ppm) of Antimony, Gold, Hafnium, Iridium, Palladium, Platinum, Rhodium, Ruthenium, and Tellurium. This is available individually as part number 04568-85.
  • Solution 7 is a 125 mL bottle of Multi-Element Solution 4 which contains 10 µg/mL (10 ppm) of Boron, Germanium, Molybdenum, Niobium, Phosphorus, Rhenium, Silicon, Sulfur, Tantalum, Titanium, Tungsten, and Zirconium. This is available individually as part number 04568-86.


Hazardous material shipping fees may apply.

Specifications
  • Purity GradeClaritas PPT® Grade
  • Concentration10 µg/mL (10 ppm)
  • ClassICP-MS
  • Element/IonSolution 1: DI Water Blank; Solution 2: Hydrochloric Acid Blank; Solution 3: Nitric Acid Blank; Solution 4: Multi-Element Solution 1; Solution 5: Multi-Element Solution 2; Solution 6: Multi-Element Solution 3; Solution 7: Multi-Element Solution 4
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionClaritas PPT® Grade ICP-MS 7 Multi-Element Solution Standards Set with Mercury
Documents & Certificates
Reviews
Q&A