Lovibond GARDNER Comparator 2000, Gardner Scale
Representative image only.
Lovibond GARDNER Comparator 2000, Gardner Scale
Lovibond | Mfr: 433340| Item: 9956240
Economical, visual color measurement in laboratory, plant, or field
£3,183.04GBP / Each
£3,819.65GBP (incl VAT)
£3,183.04GBP / Each
£3,819.65GBP (incl VAT)
Buy It Together
Current Item
Lovibond GARDNER Comparator 2000, Gardner Scale
Lovibond GARDNER Comparator 2000, Gardner Scale
9956240
£3,183.04GBP / Each
£3,819.65GBP (incl VAT)
+
Lovibond 605960 Replacement Optical Glass Cell, 10 mm
9956251
£87.49GBP / Each
£104.99GBP (incl VAT)
=
Total Price 3270.53 ADD 2 ITEMS TO CART
Product Description
Includes
Discs containing colored glass filters, sample cells/tubes, and calibration filter with certificate.
Key Features
  • Two-Field Comparator system for simple color matching
  • Supplied with lighting unit for standardized measurements
  • Color scales includes Gardner for food and beverage and colored oils, fats, and waxes
  • 1 year warranty
More About this Item
These durable, portable systems allow you to make quick visual comparisons between your sample and a range of stable colored glass standards on a disc. Battery operated illuminator offers flexibility for laboratory, plant, or field color grading. With the two-section field of view, the sample transmittance is directly compared with individual colored glass standards on a test disc.
The comparator is an easy-to-use, entry level color measurement system that compares the color of the sample directly against the color of glass in a disc. Just rotate the disc to get the closest result to the scale. Ideal for light-colored samples with visual color (pathlength up to 40 mm); an illuminator is added to systems for measuring unsaturated samples requiring a longer pathlength.
1 Year Warranty
Specifications
  • TestFood and beverage, colored oils
  • Measurement Points1 to 18 pH
  • DescriptionComparator 2000, Gardner Scale