Jenway 72 Series Simple Photometrics Measurement Video
How to Do a Simple Photometrics Measurement
Video demonstrating how to do a simple photometrics measurement on your a Jenway 7200 Series visible or UV-Visible spectrophotometer
  • Select Single Wavelength
  • SelectSimple (ABS/%T)
  • Select the wavelength to enter the required wavelength
  • Select Blank to perform a blank measurement
  • Place sample into the sammple chamber
  • Then select Read to measure the sample
Jenway 7205 Scanning UV-Visible Spectrophotometer