Industrial Filtration Basket Strainer Selection Guide
Industrial Filtration
Basket Strainer Selection Guide
Selection Considerations
  1. Amount of free open area is the ratio of the open area through the strainer basket to the cross sectional area of the pipe. A good basket strainer should have a ratio of 4 to 1.
  2. Ease of basket removal is important because baskets may require frequent cleaning or replacing due to buildup.
  3. Compactness of the design saves you valuable space. A small strainer is less bulky and easier to handle.
  4. Availability of basket perforation sizes gives the strainers the flexibility to cope with a wide range of particle sizes.