Self-Sealing Float Switch
7/14/2004
Install level-control switch without entering the tank
Cole-Parmer Instrument Company now offers a float switch that takes the headache out of float-switch installation. Just insert the switch from the outside and tighten the nut. The compression grommet forms a seal on the inside of the tank wall. Use on tanks with a wall thickness of 5/32" (4-mm) or less. Switches come in a variety of materials, including polypropylene or nylon, with nitrile or silicone grommets.
Download 300 DPI 4330104.jpg Image