Oakton® pH 600 Waterproof Handheld Meters
3/1/2006
The new Oakton waterproof, handheld pH 600-series meters feature a -2.00 to 19.99 pH range, expandable to 0.001 resolution, and accuracies up to ±0.002 pH. Units store up to 500 data sets. Time-and-date stamping meets GLP standards. Calibration capabilities include up to six calibration points, a user-settable calibration-due alarm, and up to 15 buffer options with auto buffer recognition of USA, NIST, DIN, and PWB standards. Infrared IrDA wireless technology makes PC downloading simple. IP67-rated meter operates with batteries or optional AC power adapter.
Download 300 DPI 35418_12.jpg Image