Oakton® Big-Display SaltTestr™
5/26/2005
Accurately monitor salt levels in your saline chlorination and saltwater pool systems. This rugged, new Testr takes the place of inconvenient test strips or chemical tests. Simultaneous readout of temperature and salt level in ppt appears in a full 1-1/16"H display. IP67-rated waterproof housing is designed to float!
Download 300 DPI 3566151.jpg Image