Low-cost, reliable stirring and heating
Cole-Parmer introduces a new line of digital stirrers, hot plates, and stirring hot plates designed for general laboratory applications.
Units feature a two-piece spill-resistant aluminum housing to deflect spills away from internal components.Microprocessor-controlled speed and temperature can be precisely controlled throughout the entire range. Stirrers’ “instant ON” feature starts stirring at 50 rpm to minimize splashing. Top plates are available in a corrosion-resistant ceramic for high-temperature applications or rugged aluminum for a more uniform temperature.
Advanced hot plate and stirring hot plate are designed for applications that require more precise accuracy and repeatability. Units feature an updated safety system which includes a hot surface warning indicator and over-temperature protection. Individual heating and stirring displays, timer, and external temperature probe kit ensure repeatable results.