New PVC Solvent Autoanalysis Tubing
9/13/2004
Low-cost choice for solvent transfer
Cole-Parmer Instrument Company introduces a new, low-cost PVC Solvent Autoanalysis tubing ideal for transferring aggressive solvents in chromatography applications or peristaltic pump transfer. This translucent yellow tubing allows good visibility of the fluid passing through. Tubing is available with either two or three stops in sizes ranging from 0.19 to 2.79 mm ID.
Download 300 DPI 9560610.jpg Image