New Precision Turbine Flowmeters
11/14/2007
Highest accuracy with full-featured electronics
Cole-Parmer introduces a new line of precision turbine flowmeters for medium to large batching of water or chemicals, for valve and switch control in tank-filling for municipalities, or for other applications where accuracy and control is a necessity. Using the turbine measurement principle, these meters are ideal for applications involving higher viscosity and aggressive fluids.
Precision turbine flowmeters are available with or without display. Models with display feature a 6-digit LCD with 1/2″H characters. Choose no output, 4 to 20 mA, or scaled-pulse output. Input power is 8.5 to 35 VDC. All meters feature the “easiest in the industry” push-button programming and feature an electronic lockout to ensure against tampering or resetting.
Cole-Parmer offers a 3/4″ to 2″ sanitary process connection unit or a 1/2″ to 3″ NPT(M) process connection unit. Meters can handle particulate sizes from 420 to 1410 µm, depending on process connection size.