Eliminate the extra time needed to order individual parts, assemble components and test a water treatment system
Cole-Parmer Instrument Company introduces a new line of Cole-Parmer cooling tower control systems for hassle-free solutions to water treatment needs. Customize a system to fit your specific needs or select from one of four standard systems: basic conductivity system; basic ORP and conductivity system; basic pH and conductivity system; or advanced pH, ORP, and conductivity system.
Place these systems in almost any location. Simply attach the PVC back plane with four bolts, join the inlet/outlet and chemical connections, and plug into a power outlet. Each system is pressure tested to 100 psi for quality assurance. Broaden a system's capabilities by adding options such as corrosion coupon racks, flowmeters, datalogging interfaces, and tanks.
Download 300 DPI 7078900.jpg Image