NEW Oakton® Waterproof EcoTestr™ pH 2 Pocket Meter
9/1/2008
Easy-to-read pH measurement with an easy-to-afford price!
The latest addition to the OAKTON pHTestr® family features an ergonomically designed LCD, allowing you to simply dip and read without having to turn your head or tilt the tester. The meter’s rugged body is both waterproof and dustproof for use in dirty and damp conditions, and even floats if dropped in water. Its budget-friendly price makes it great for environmental or educational applications where meters may become lost or damaged.
The EcoTestr pH 2 features a long-life pH sensor that is recessed to protect against breakage, automatic buffer recognition for quick and easy one- to three-point calibrations, and automatic temperature compensation for accurate reading under changing conditions.