NEW Oakton® Mini TempTestr® IR Thermometer
4/20/2004
Pocket-sized and economical- measure temperatures as low as -33°F and as high as 430°F!
Cole-Parmer Instrument Company introduces the Oakton Mini TempTestr IR, a non-contact infrared thermometer that takes quick, accurate readings, yet-at 1-½"W x 2-½"H x ½"D-is small enough to fit in your pocket.
The Mini TempTestr IR thermometer (39642-00) will measure the surface temperature of any liquid, solid, or semi-solid in less than one second with an accuracy of up to ± 1.1°F (0.6°C). That makes it ideal for heating and air conditioning, water quality, industrial, construction, electrical, horticulture, and a variety of other applications.
The Mini TempTestr IR features a minimum/maximum memory reading, Hold function, continuous-on setting, adjustable emissivity, flip-open stand, easy-to-read display, and soft touch keypad. It comes with a wrist strap, metal storage case, and extra battery.
Download 300 DPI 3964200.jpg Image