Exceptionally bright laser target is visible even in sunny areas!
Cole-Parmer Instrument Company proudly introduces the newly redesigned Oakton InfraPro series of infrared thermometers. These units provide non-contact temperature measurement and feature an exceptionally bright laser targeting system that is visible even in sunny areas.
The new InfraPro series is available in four models: from the InfraPro 1, which measures temperatures up to 750°F (400°C) and has a distance-to-spot ratio of 12:1, to the highly precise InfraPro 4, which measures temperatures up to 1400°F (760°C) and offers a distance-to-spot ratio of 50:1. An intrinsically safe model is also available for use in hazardous environments.
Typical applications include measuring food and beverages, checking heating/cooling ducts and registers, checking agricultural plants, and monitoring steam and motor temperatures.