NEW Oakton® DO 6 Dissolved Oxygen Meter
5/7/2004
Quality and convenience at an economical price
Cole-Parmer Instrument Company introduces the new DO 6 handheld dissolved oxygen meter from Oakton Instruments. This pocket-sized unit builds upon a convenient, compact body design to combine high accuracy with economical pricing.
Test water quality from a wide variety of sources with measuring ranges of 0 to 20 ppm or 0 to 200% saturation. Six buttons allow for simple setup, calibration, display values, and electrode diagnostic capability. Manually input the barometric pressure and salinity values for automatic compensation of readings.
The meter includes a low-maintenance, Galvanic probe so that no warm-up time is required. Membrane replacement is easy with the cap system-just twist off worn membrane, fill with solution, and replace with new membrane cap. Additional features include a three-year warranty on the meter, and a protective rubber boot that doubles as a benchtop stand. CE-approved.
Download 300 DPI 3564210.jpg Image