NEW Oakton® Basic pH 11 and Advanced pH 110 pH/mV/Temperature Meters
4/26/2004
Memory increased to 50 and 100 data sets; calibration to five and six pH points respectively
Cole-Parmer Instrument Company introduces the NEW Oakton pH 11 and pH 110 meters, the latest product upgrades in our popular Oakton line of handheld pH meters.
Meter enhancements include a large, dual LCD that lets you view pH (or mV) and temperature values simultaneously, and automatic temperature compensation (ATC) to ensure accuracy even in fluctuating temperatures. Functions include temperature offset calibration, Hold, ready indicator, and Auto-Off. Decipherable icons and electrode slope display capability allow quick troubleshooting.
The Basic pH 11 meter offers calibration for up to five pH points from USA and NIST buffers, and memory for up to 50 data sets. The Advanced pH 110 meter adds relative mV offset calibration and reading capabilities, calibration for up to six pH points from USA, NIST, DIN, and PWB buffer options, and nonvolatile memory for up to 100 sets of data. The pH 110 also includes free software and a RS-232 output for downloading data to a printer or computer. Print all or select data continuously, or in selected time intervals between 10 and 24 seconds.
Download 300 DPI 3561520.jpg Image