NEW Masterflex® L/S® Multichannel Pump Heads
4/14/2004
Up to 32 synchronous pumping channels without cartridges
Cole-Parmer introduces six new models of Masterflex L/S multichannel pump heads-the 07534-, 07535-, and 07536-series-which are designed to deliver the efficiency and versatility of synchronous flow rates from up to 32 separate channels.
The flow range for these multichannel heads is 0.0026 to 2300 mL/min per channel, depending on drive rpm and tubing selection. They accept several formulations of two-stop Masterflexr tube sets in microbore and L/S precision and L/S high-performance precision sizes. The tubing is quick and easy to load, and stays firmly in place. Simply switch tubing to alter the flow range or to accommodate different fluids.
These durable, continuous-duty pump heads are available with six rollers (07534- and 07535-series) for low-pulsation flow, or with three rollers (07536-series) for maximum flow. The heads feature a frame made of rugged anodized aluminum complimented by stainless steel rotor/roller assemblies and shielded, stainless steel bearings. The precision-machined aluminum occlusion beds apply optimum compression to the tubing so that no occlusion adjustment or cartridges are required.
Multichannel pump heads are available individually or they can be purchased as part of a system that includes an L/S variable-speed drive and Masterflex L/S tubing.
Download 300 DPI 7792700.jpg Image