Vernon Hills, Ill—The best of the globally tried, tested and trusted technology of Masterflex combined with the Swiss precision engineering of Ismatec offers pump operators a pumping experience with the NEW Masterflex® Ismatec® Reglo Digital Peristaltic Pump Series. Over 100 years of excellence has been drawn together, culminating in the first major design refresh for the Reglo Peristaltic Pumps. These devices truly deliver powerful performance with intuitive interfacing, making it easier than ever to achieve the results operators need.
Masterflex Ismatec Reglo Digital peristaltic pumps are ideal for the laboratory, with flow-driven technology that supports precision dispensing; functional versatility giving freedom in applications; and a space-saving compact footprint to conserve valuable benchtop space. In comparison to its predecessor, the new Reglo pumps have a wider overall flow range with the option of dosing between 0.00009- and 368-mL/min. The models pack in advanced features designed to deliver fluids accurately, including anti-drip technology to help conserve valuable samples, speed ramping to better accommodate fluids with varying viscosities, and touch-screen interfacing that supports powerful programmable performance—all while taking up very little space (178 x 152 x 165; H x L x W, mm), allowing these pumps to be integrated into even the most space-limited areas.
The cartridge design synonymous with Ismatec Reglo pumps has been retained, with a choice of two or four channels, both of which fit three-stop tubing. The cartridge configurations offer a choice of 6, 8 and 12 rollers, allowing you to choose the optimal balance of flow rate and pulsation. For the first time in the Reglo family of pumps, the popular Miniflex® pump heads have been integrated into the available options, giving you a choice of one- or two-channel configurations, both of which use continuous L/S® tubing. Furthermore, these new Reglo pumps feature a 50W BLDC motor instead of the 75W PMDC motor used in older models, allowing for powerful yet efficient performance. These small but mighty pumps continue the Ismatec legacy of being ideal for feed, transfer, perfusion, volume dispensing and time dispensing.
As hinted at above, these new Reglo pumps offer an easy-to-navigate and intuitive 5” capacitive touchscreen display so it can be used with disposable gloves. The Android™ platform on which the interface is designed helps the menu structure feel instantly familiar to most users. In addition, these are the first highly accurate micro-flow (<100-mL/min) models to receive MasterflexLive™ capability and 21 CFR Part 11 and EU Annex 11 compliancy. This secure, Cloud-based platform allows you to control and monitor your pump via a PC, tablet or smartphone (iOS™ and Android™). You can ensure that valuable sample loss is minimized, and your safety is optimized, due to the ability to control of all pump parameters, including speed, flow rate, dispense volume, and more in real-time and remotely. Push notifications provide alerts for operating conditions and error messages. By subscribing to MasterflexLive premium, gain access to 21 CRF Part 11 and EU Annex 11 data compliance with the ability to robustly manage data, maintain digital records and control access to data.
For more information, go to www.coleparmer.com/masterflex or www.masterflex.com.
Popular Categories
- Balances and Scales
- Analytical Balances
- Bench and Floor Scale Bases
- Bench and Floor Scales
- Counting Scales
- Crane and Hanging Scales
- Health Scales
- Indicators for Scales
- Mechanical Balances
- Moisture Balances
- Platform and Pallet Scales
- Production Weighing
- Semi and Micro Analytical Balances
- Toploading Balances
- Weighing Accessories
- Weighing Dishes
- Calibration
- Chemicals
- Electrical Products
- Batteries and Battery Testers
- Cable Testers
- Cable Ties
- Clamp Meters
- Electrical Accessories
- Electronics Enclosures
- Function Generators
- Gauss Meters
- Insulation Testers
- LCR Meters
- Megohmmeters
- Motor Rotation Testers
- Multimeters
- Oscilloscopes
- Power Meters and Analyzers
- Power Supplies
- Resistance Testers
- Surge Protectors
- Test Leads and Probes
- Voltage Detectors
- Filtration Products
- Flow, Level and Valves
- Fluid Handling
- Furniture
- Heating and Cooling Equipment
- Laboratory Equipment
- Baths and Circulators
- Burettes (Burets)
- Calorimeters
- Centrifuges
- Chromatography Products
- Desiccators
- Digestion Systems
- Dispensers
- Distillation
- Electrophoresis Products
- Electroporation Products
- Environmental Samplers
- Extraction Apparatus
- Fermentation Products
- Flame Photometers
- Fluid Bed Dryers
- Freeze Dryers
- Glassware Washers
- Histology Products
- Homogenizers and Ultrasonic Processors
- Humidifiers and Dehumidifiers
- Lab Shakers
- Laboratory Evaporators
- Laboratory Sieves and Sieve Shakers
- Leachate Equipment
- Lighting and Illumination
- Magnetic Stirrers and Hot Plates
- Microscopes and Magnification
- Mixers and Blenders
- Particle Counters
- Pipettes and Pipettors
- Refractometers
- Thermal Cyclers
- Titrators
- Ultrasonic Cleaners
- Vacuums
- Water Purification Products
- Laboratory Supplies
- Air Sampling
- Animal Research Products
- Bags
- Beakers
- Books and Reference Materials
- Bottles and Jars
- Calculators
- Carboys
- Chemistry Kits
- Clean Room Stationery
- Containers
- Cuvettes
- Dialysis Products
- Dippers
- Distillation Supplies
- Field Sampling
- Flasks
- Graduated Cylinders
- Ice Buckets and Ice Pans
- Lab Cleaning Products
- Lab Organizers
- Labels and Label Printers
- Laboratory Dishes
- Laboratory Timers and Clocks
- Labware
- Microbiology Products
- Microplate Products
- Mortars and Pestles
- Parts and Accessories
- Racks
- Sampling Wipes
- Shippers
- Syringes
- Tubes
- Vials
- Watch Glasses
- Material Handling and Storage
- Material Testing and Physical Testing
- Pressure and Vacuum Instruments
- Process Controllers
- Pumps
- Safety Products
- Air Quality Monitors
- Arc Flash Products
- Combustion Analyzers
- Eyewash
- Eyewear and Safety Glasses
- Floor Mats and Matting
- Gas Detectors
- Gloves
- Hearing Protection
- Lockout and Tagout Product
- Radiation Detectors
- Refrigerant Products
- Respirators and Masks
- Safety Apparel
- Safety Shields
- Safety Signs and Labels
- Solvent Safety
- Sound Meters
- Spill Control
- Temperature Measurement Products
- Test and Measurement Products
- Water Quality Products
- Colorimeters
- Colorimetry Reagents
- Conductivity, Resistivity, and TDS Meters
- Dissolved Oxygen Meters
- Dye Tracers
- Electrodes
- Flocculation Testers
- Multiparameter Meters
- pH Buffers
- pH Meters
- Turbidity Accessories
- Turbidity Meters
- Water Quality Accessories
- Water Quality Standards, and Solutions
- Water Quality Test Strips