New Masterflex B/T Process Tubing Pumps - Improved Housing!
9/7/2006
Improved Housing! Improved Tubing! Improved Loading!
Cole-Parmer introduces the new and improved line of Masterflex® B/T® process pumps featuring a redesigned, intuitive loading system for quick and easy tubing changes.
At the heart of the redesign is a new occlusion mechanism that slides the tubing bed out of the way, and a retention mechanism that grips the tubing for trouble-free loading. Additionally, the easy-clasp closure provides one-handed opening and locks closed without the use of tools.
Performance of these pumps has been improved through the use of new PerfectPosition™ tubing, which features retention marks to indicate the exact tubing alignment needed for optimal performance and tubing life. Two new tubing sizes—B/T 87 and B/T 91— provide enhanced pumping performance in high-pressure applications.
In addition, an improved, cast-aluminum housing offers quieter operation while withstanding rugged conditions. An epoxy-powder coating on the housing prevents chemical corrosion and improves cleanup. The easy-open front cover swings away for CIP or SIP protocols.