New Line of Digi-Sense® Thermocouple Wire
9/9/2004
Features high-temperature insulation up to 2,600°F (1,427°C)!
Cole-Parmer Instrument Company introduces a new line of Digi-Sense thermocouple wire designed to be used in the manufacturing of temperature sensors and extension cables.
This new thermocouple wire is constructed of calibrated copper conductors for high system accuracy. Choose from a selection of insulation materials for the most demanding applications. Ceramic fiber braid insulation is designed for high-temperature applications up to 2600°F (1427°C). Fiberglass braid insulation is non-flammable and offers superior abrasion resistance. FEP insulation features excellent durability and mechanical strength, and resists most chemicals. Or, select our economically priced PVC insulation for general-purpose use. Available in a variety of wire gauges, insulation materials and spool lengths.
Ideal for industrial, research, pharmaceutical, and aerospace applications. Developed using the latest technology, this wire is subjected to a lengthy series of electrical, mechanical, and chemical tests-one of the most rigorous quality assurance programs in the thermal instrumentation industry.
Download 300 DPI 0854100_se.jpg Image