New from Cole-Parmer: Ismatec Reglo Independent Channel Control Peristaltic Pump
7/23/2013
Independently control flow and direction of each channel in a single compact unit
Vernon Hills, Ill., July 23, 2013—Expand benchtop pumping capabilities with the new Ismatec® Reglo Independent Channel Control Peristaltic Pump from Cole-Parmer. This three-channel peristaltic pump lets users individually control the flow and direction of each channel from the intuitive keypad or via computer. They can also make fast changeovers with the easy-to-use tubing cassettes. A USB interface offers quick connections.
The micro peristaltic pump delivers continuous pumping or precision dispensing up to 35 mL/min with the capability of bidirectional flow in each channel. Independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy.
To view the Ismatec® Reglo Independent Channel Control Peristaltic Pump, go to ColeParmer.com/20256 or call 800-323-4340 for more information.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at www.ColeParmer.com
Download 300 DPI 78001_70.jpg Image