New, Flexible Fluoro-Elastomer Tubing
7/14/2004
The clear choice for chemically aggressive applications
Cole-Parmer Instrument Company introduces the revolutionary, new Thermoflon high-performance tubing, which combines the chemical-resistance of fluoro-plastics with the flexibility of fluoro-rubbers.
This fluoro-elastomer tubing has a durometer of 67(A), and a bend radius as small as 1/2"a flexibility that makes fitting selection easy and prevents tubing from kinking.
Tubing is available in three variations. Thermoflon Basic is a single-layer tubing with the highest level of flexibility and the lowest price. Thermoflon Advanced is a dual-layer tubing with improved chemical resistance and less potential for gas permeability. Thermoflon Pascal is a dual-layer tubing with the highest level of chemical resistance and pressure durability, and the lowest level of solution permeability.
All three Thermoflon grades are chemical-resistant (even to highly oxidative substances such as ozone), and will not leach chemical additives. The tubing is also clear, allowing users to visually monitor the contents.
Download 300 DPI 9562500.jpg Image