Inner coating prevents microorganism growth
Cole-Parmer introduces a revolutionary, new anti-microbial tubing, ideal for applications where microorganism contamination is a concern.
A coating of silver ions on the inner wall of the tubing protects it from bacterial, mold, and fungi growth. Because of this antimicrobial protection, the user can replace the tubing less frequently. The coating remains effective over time and is not temperature sensitive. This tubing is translucent blue, prpoviding good fluid visibility. Compatible with standard barbed fittings.
The silver ions will not contaminate the fluid, making Flexelene Silver tubing ideal for general laboratory, food/beverage, pharmaceutical, and biotech applications.
Download 300 DPI 9617000.jpg Image