Select from an easy-to-use humidity meter or a meter with datalogging capabilities
Cole-Parmer Instrument Company introduces the latest Digi-Sense meters for temperature, humidity, and dew point measurement-available as an easy-to-use humidity meter or a meter with datalogging capabilities.
These thermohygrometers simultaneously display temperature, humidity, and dew point, eliminating the need to switch screen displays. The rugged IP54-rated design allows measurements in industrial applications, and offers splash- and dust-resistant protection. Single- or dual-point temperature and humidity calibration provides convenient field accuracy checks.
The easy-to-use, low-cost thermohygrometer (37003-10) features selectable °F/°C temperature measurement, screen hold, low-battery indicator, and a built-in stand. The datalogging thermohygrometer (37003-02) features all of the above, plus the capability to log up to 1,000 time- and date-stamped measurements, programmable logging interval from one second to 60 minutes, RS-232 data download or infrared signal output, adjustable filter rate (stabilize fluctuating readings), min/max readings, and recall function.
Both units measure humidity with an accuracy of ±2% and generate temperature readings accurate to ±1°F (±0.5°C).
Download 300 DPI 3700310.jpg Image