Cole-Parmer Instrument Company introduces the new Digi-Sense palm-sized infrared thermometer with Class II laser sighting for measuring and displaying noncontact temperatures. Thermometer fits perfectly in your hand or pocket. Measures temperatures as high as 482
Unit features a distance-to-target ratio of 8:1, with an adjustable emissivity range of 0.5 to 1 for a wider selection of surface measurements. The LCD simultaneously displays the current temperature (
Download 300 DPI 3964300.jpg Image