NEW Cole-Parmer® Symmetry™ P-Series Balances
3/29/2006
Precision toploading and analytical balances
The Symmetry P Series (PR and PA) features precision toploading and analytical balances. Designed for precision work, these balances feature multiple weighing units and functions used in most laboratories, RS-232 interface for GLP compliant record keeping, and durable aluminum housing. PR precision toploading models offer capacities ranging from 210 to 4400 g, and readability from 0.001 to 0.01 g. PA precision analytical units offer capacities of 120 and 220 g and readability of 0.0001 g. these balances are ideal for the pharmaceutical industry and labs.
Download 300 DPI 10000_12left.jpg Image