New Cole-Parmer® Symmetry® Balances
2/5/2008
Economical toploading balances for a wide variety of applications
Cole-Parmer introduces the second-generation Symmetry EC II balance with extra features for the same low price.
These upgraded Symmetry EC II models now have a standard draft shield, security lock-down port, and an internal rechargeable battery for improved weighing convenience. All balances have capacities of 120 to 4000 g with readability of 0.005 to 0.1 g. Each model features a capacity tracker to warn of approaching overload and a RE-232 interface for connection to PCW printer.
Completing the Symmetry E Series line, our ED models offer quality solutions for educational institutions seeking a basic, low-cost balance. Each ED balance has readability ranges from 0.01 to 1.0 g and capacities from 150 to 4000 g.