New Cole-Parmer®-Brand Plasticware
9/28/2006
Cole-Parmer introduces a new line of proprietary plasticware that includes narrow- and wide-mouth bottles, wash bottles, and beakers, economically bulk-packed for greater cost savings.
Bottles are offered in three different materials—LDPE, HDPE, and PP—with leakproof PP enclosures included. Translucent LDPE bottles are ideal for food and general laboratory use. HDPE bottles are manufactured with FDA-approved resin for noncontaminating storage of chemicals and food-grade products. Rigid PP bottles offer superior chemical resistance and are ideal for storage and sampling of liquids and solids.
Wash bottles are manufactured of LDPE and feature a built-in dispenser that gives an ultrafine stream. Contents can be dispensed without tilting the bottle and the wide mouth allows for faster, easier filling.
Griffin-style graduated beakers are manufactured of PP and offer superior chemical resistance. They can withstand temperatures to 275°F (135°C).