Masterflex® PharmaPure® Tubing
1/8/2007
Ultra-low particle spallation for sterile filling applications
Cole-Parmer offers PharmaPure, an exciting new tubing formulation for the MASTERFLEX peristaltic pump series. This co-extruded tubing has an inner liner that virtually eliminates particulate spallation into the fluid, and a soft, flexible outer layer that permits use in a peristaltic pump.
PharmaPure not only maintains the sterility of the fluid, it provides superior flex life, excellent wear properties, and low permeability. In addition, it meets all USP Class VI and FDA criteria.
PharmaPure 06435-series tubing is available in the MASTERFLEX L/S®, I/P®, and B/T® sizes. Like other MASTERFLEX tubing formulations, PharmaPure is manufactured to extremely close tolerances, ensuring accurate and repeatable flow.